BUFFER SOLUTION PH 6.87 25C 1 * 500 mL
Catalog # REGEEP687
Supplier: REAGECON
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Polycarbonate erlenmeyer flasks with screw caps, Corning®
Supplier: Corning
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
BUFFER SOLUTION PH 4.01 25C 1 * 500 mL
Supplier: REAGECON