ContactUsLinkComponent
Box, storage, stackable, neobox-100, material PP, for vials, diameter 12mm, transparent, autoclavable, tight-closing, transparent hi nged lid, base with different colors, width 140mm, height 153mm, depth 55mm. 1 * 1 items
Catalog # KOEH2-1967
Box, storage, stackable, neobox-100, material PP, for vials, diameter 12mm, transparent, autoclavable, tight-closing, transparent hi nged lid, base with different colors, width 140mm, height 153mm, depth 55mm. 1 * 1 items
Catalog # KOEH2-1967
Frequently Bought Together