Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Box of 96 PhyTip columns (300 µL volume) containing 20 µL of Normal Phase I resin per column
Catalog # PHYNPTH93-20-09
Supplier:
PHYNEXUS, INC.
Expand image
Box of 96 PhyTip columns (300 µL volume) containing 20 µL of Normal Phase I resin per column
Catalog # PHYNPTH93-20-09
Supplier:
PHYNEXUS, INC.