Buffer Solutions
13,00, ±0.01, 25 °C, 4 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Dokumente
Certificates
Search by Lot Numbermsds_ALFA038712 msds_ALFA038714 msds_ALFA040439 msds_ALFA040447 msds_ALFA042416
Sicherheitsdatenblatt
More
Specifications
Specifications
- Pk:4 L
- Reference temperature:25 °C
- pH accuracy:±0.01
- pH value:13,00
- Cat. No.:46307.K7
- Physical form:Liquid
- MDL No.:MFCD03458582
- Appearance:Colourless
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



