Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer solution pH 1,09

Buffer solution, 1,09 (25 °C), 1 L
Catalog # KRAF03853.3060
Supplier:   Bernd Kraft
Buffer solution pH 1,09
Catalog # KRAF03853.3060
Supplier:  Bernd Kraft
CAS Number:  
Buffer solution pH 1,09
Buffer solution pH 1,09

Buffer solution, 1,09 (25 °C), 1 L

CAS Number:  
Price
64,40 €
Log in for availability
1 * 1 L
Product Details & Documents
  • Stabilized

Certifications: According to DIN 19267.

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Buffer solution
  • pH value:
    1,09 (25 °C)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.