Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Laboratory jacks

200×230 mm, 60 kg, 7 kg, 70 - 260 mm, Blue
  JUCHVL02291/3
Laboratory jacks
  JUCHVL02291/3
Laboratory jacks
Laboratory jacks

200×230 mm, 60 kg, 7 kg, 70 - 260 mm, Blue

 
152,20 €
   
1 * 1 ST
 

Aluminium alloy, with epoxy resin coating. These manually operated lifting stages provide high stablity.

  • Special epoxy resin coating ensures that the surfaces are extremely resistant to chemical attack and to other environmental factors
  • Strict quality controls, as required by DIN EN ISO 9001:2000, ensure a constant product quality
  • Made in Germany
 
  • Max. static load capacity:
    60 kg
  • Plate size:
    200×230 mm
  • Working height:
    70 - 260 mm
  • Colour:
    Blue
  • Seq:
    06
  • Max. dynamic load capacity:
    7 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.