Order Entry
Germany
Orders LinkContactUsLinkComponent
Adapters, sperical joints

Adapters, sperical joints

Socket S 35 with steel thread M 33×1,5, O.Ø 30×2,4 mm, I.Ø 25,2 mm
Catalog # JUCH20457.35
Adapters, sperical joints
Catalog # JUCH20457.35
Adapters, sperical joints
Adapters, sperical joints

Socket S 35 with steel thread M 33×1,5, O.Ø 30×2,4 mm, I.Ø 25,2 mm

Price
728,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

For reaction vessels, 316 stainless steel, thread length 14 mm.

Specifications
  • Type:
    Socket S 35 with steel thread M 33×1,5, O.Ø 30×2,4 mm, I.Ø 25,2 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.