Order Entry
Germany
Orders LinkContactUsLinkComponent
dNTP mix

dNTP mix

dNTP mix, 10 mm each, size: 5×1 ml, 5
Catalog # 40054.
Supplier:   Biotium
dNTP mix
Catalog # 40054.
Supplier:  Biotium
CAS Number:  
dNTP mix
dNTP mix

dNTP mix, 10 mm each, size: 5×1 ml, 5

CAS Number:  
Price
482,00 €
Log in for availability
1 * 5 ST
Product Details & Documents

Highly purified mix of dNTPs (>99% by HPLC) in 0,6 mM Tris-HCl (pH 7,5); each dNTP is at a concentration of 10 or 25 mm in the solution provided.

The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.

Specifications

Specifications

  • Pk:
    5
  • Description:
    dNTP mix, 10 mm each, size: 5×1 ml

Specifications