Distillation flask, STANHOPE-SETA
500 ml
Catalog # STAN11910-001
Supplier: STANHOPE SETA
Jump to:
Product Details & Documents
For the distillation of petroleum products.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Complies with ASTM D850.
Specifications
- Kapazität:500 ml
- Seq:02