Order Entry
Germany
Orders LinkContactUsLinkComponent
Dipping Pipettes

Dipping Pipettes

Dipping pipette, tolerance ±0,1 ml, 50 ml
Catalog # NEUB0276-50-050
Dipping Pipettes
Catalog # NEUB0276-50-050
Dipping Pipettes

Dipping pipette, tolerance ±0,1 ml, 50 ml

Price
70,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Dipping pipettes for sampling predefined volumes. The pipettes are immersed into the samples and filled via the open top. The pipette is emptied by tilting sideways. Calibrated to deliver EX.

Specifications
  • Capacity:
    50 ml
  • Description:
    Dipping pipette, tolerance ±0,1 ml
  • Seq:
    04