Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Capillary columns, TRACE™ TR-5MS

Capillary, TR-5MS, 0,18 µm, 0,18 mm, 20 m
  555-9497
Capillary columns, TRACE™ TR-5MS
  555-9497
Capillary columns, TRACE™ TR-5MS
Capillary columns, TRACE™ TR-5MS

Capillary, TR-5MS, 0,18 µm, 0,18 mm, 20 m

 
719,00 €
   
1 * 1 ST
 

TRACE™ TR-5MS GC columns are ideal for hydrocarbons, solvents, pesticides, herbicides, phenols, amines.

  • Non-polar phase, 5% phenyl polysilphenylene-siloxane
  • Low bleed and high stability
  • High signal-to-noise for increased sensitivity
  • High robustness to oxygen and water contamination
 
  • Column Format:
    Capillary
  • Film thickness:
    0,18 µm
  • Ø int.:
    0,18 mm
  • Length:
    20 m
  • Phase:
    TR-5MS