Order Entry
Germany
Orders LinkContactUsLinkComponent
Capillary columns, TRACE™ TR-FFAP

Capillary columns, TRACE™ TR-FFAP

Capillary, TR-FFAP, 0,50 µm, 0,32 mm, 50 m
Catalog # 555-9406
Capillary columns, TRACE™ TR-FFAP
Catalog # 555-9406
Capillary columns, TRACE™ TR-FFAP
Capillary columns, TRACE™ TR-FFAP

Capillary, TR-FFAP, 0,50 µm, 0,32 mm, 50 m

Price
1 370,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

TRACE™ TR-FFAP GC columns produce outstanding robustness in difficult-to-analyse separations.

  • Engineered specifically for FFAP analysis
  • Polar phase, TPA modified polyethylene glycol
  • Quality tested for acidic compound analysis
  • Equivalent to USP G25 and G35
  • Excellent quality and reproducibility
  • Low bleed even at elevated temperatures
Specifications
  • Column Format:
    Capillary
  • Film thickness:
    0,50 µm
  • Ø int.:
    0,32 mm
  • Length:
    50 m
  • Phase:
    TR-FFAP