Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Hydroxybenzhydrazide

4-Hydroxybenzhydrazide

Glass bottle for solids, 25 g
Catalog # FLUO018715-25G
Irritant
Marketsource
Supplier:   FLUOROCHEM
4-Hydroxybenzhydrazide
Catalog # FLUO018715-25G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  5351-23-5
4-Hydroxybenzhydrazide

Glass bottle for solids, 25 g

CAS Number:  5351-23-5
Price
57,10 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  4-Hydroxybenzoic hydrazide, 4-Hydroxybenzohydrazide, p-Hydroxybenzoic acid hydrazide, 4-Hydroxybenzene-1-carbohydrazide, 4-Hydroxybenzoic acid hydrazide
  • CAS Number:5351-23-5
  • Formula:C₇H₈N₂O₂
  • MDL Number:MFCD00007605
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:370 °C (1013 hPa)
  • Melting Pt:266…269 °C
  • Storage Temperature:Ambient
  • EINECS:226-326-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.