Order Entry
Germany
Orders LinkContactUsLinkComponent
5'-Bromo-2'-hydroxypropiophenone

5'-Bromo-2'-hydroxypropiophenone

5 g
Catalog # FLUO012088-5G
Marketsource
Supplier:   FLUOROCHEM
5'-Bromo-2'-hydroxypropiophenone
Catalog # FLUO012088-5G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  17764-93-1
5'-Bromo-2'-hydroxypropiophenone

5 g

CAS Number:  17764-93-1
Price
144,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  1-(5-Bromo-2-hydroxyphenyl)propan-1-one
  • CAS Number:17764-93-1
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD00218577
  • Molecular Weight:229.0706 g/mol
Specifications

Specifications

  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.