Order Entry
Germany
Orders LinkContactUsLinkComponent
 

5'-Bromo-2'-hydroxypropiophenone

25 g
Catalog # FLUO012088-25G
Marketsource
Supplier:   FLUOROCHEM
5'-Bromo-2'-hydroxypropiophenone
Catalog # FLUO012088-25G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  17764-93-1
5'-Bromo-2'-hydroxypropiophenone

25 g

CAS Number:  17764-93-1
Price
654,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1-(5-Bromo-2-hydroxyphenyl)propan-1-one
  • CAS Number:17764-93-1
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD00218577
  • Molecular Weight:229.0706 g/mol
Specifications

Specifications

  • Pk:
    25 g

Specifications