4-(Boc-amino)cyclohexanone
Glass bottle for solids, 1 g
Catalog # FLUO011548-1G
Marketsource
Supplier: FLUOROCHEM
4-(Boc-amino)cyclohexanone
Catalog # FLUO011548-1G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 179321-49-4
Jump to:
Product Details & Documents
Synonyms: 4-Aminocyclohexan-1-one, N-BOC protected, tert-Butyl (4-oxocyclohexyl)carbamate
- CAS Number:179321-49-4
- Formula:C₁₁H₁₉NO₃
- MDL Number:MFCD00798168
- Molecular Weight:213.2700 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



