Extender tools
Extender tool, Aluminium, Standard knurl
Catalog # UPCHP-299
Supplier: UPCHURCH SCIENTIFIC
Jump to:
Product Details & Documents
These tools made from aluminium or Delrin® can be used to tighten most knurled nuts in hard to reach places. For precise tightening, the extender tools listed with ¼" hex drives are designed to adapt to any torque wrench with a female ¼" socket.
Specifications
- Description:Extender tool
- Material:Aluminium
- Use with head style:Standard knurl