Orders LinkContactUsLinkComponent
Extender tools
Extender tools
Extender tool, Aluminium, Standard knurl, short
Catalog # UPCHP-399
Supplier:   UPCHURCH SCIENTIFIC
Extender tools
Catalog # UPCHP-399
Extender tools
Extender tools
Extender tool, Aluminium, Standard knurl, short
Price
42,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
These tools made from aluminium or Delrin® can be used to tighten most knurled nuts in hard to reach places. For precise tightening, the extender tools listed with ¼" hex drives are designed to adapt to any torque wrench with a female ¼" socket.
Specifications
  • Description:
    Extender tool
  • Material:
    Aluminium
  • Use with head style:
    Standard knurl, short