Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Storage Bins

Blue, 230×150×125 mm, PP
Catalog # SEMA2796
Supplier:   SEMADENI
Storage Bins
Catalog # SEMA2796
Supplier:  SEMADENI
Storage Bins
Storage Bins

Blue, 230×150×125 mm, PP

Price
12,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Made of PP.

  • Secure stacking
  • Made of high quality material and are dimensionally stable
  • Supporting edge and smooth inner surfaces for easy cleaning

Storage bins with large labelling field for content description.

Specifications
  • Colour:
    Blue
  • Material:
    PP
  • Seq:
    03
  • W×D×H:
    230×150×125 mm