Electrode Cables
Electrode cable, 2×2 mm, for connecting temperature sensors with Metrohm plug-in head G with 780, 781, 826, 827 pH/Ion meters and all Titrandos, 1 m
Catalog # METR6.2104.140
Supplier: METROHM
Jump to:
Product Details & Documents
Specifications
- Description:Electrode cable, 2×2 mm, for connecting temperature sensors with Metrohm plug-in head G with 780, 781, 826, 827 pH/Ion meters and all Titrandos
- Length:1 m
- Seq:09