ContactUsLinkComponent
HP TOTAL RNA KIT (C-LINE) PEQGOLD 200 Preps Isolation of RNA from fatty tissues. Yield: up to 100 µg. Kit Components: PerfecBind RNA Columns (Classic-Line), Collection Tubes, TriFast Reagenz, RNA Wash Buffer I and II, RNase-free Water Shipment: RT St 1 * 200 Tests
Catalog # PEQL12-6936-02
Supplier:  VWR Peqlab
HP TOTAL RNA KIT (C-LINE) PEQGOLD 200 Preps Isolation of RNA from fatty tissues. Yield: up to 100 µg. Kit Components: PerfecBind RNA Columns (Classic-Line), Collection Tubes, TriFast Reagenz, RNA Wash Buffer I and II, RNase-free Water Shipment: RT St 1 * 200 Tests
Catalog # PEQL12-6936-02
Supplier:  VWR Peqlab
Related Information