Order Entry
Germany
Orders LinkContactUsLinkComponent
Friedman-Pearson rongeurs

Friedman-Pearson rongeurs

Friedman-Pearson rongeurs, 0,7 mm cup diameter, Curved, 140 mm
Catalog # FSCI16121-14
Marketsource
Supplier:   FINE SCIENCE TOOLS
Friedman-Pearson rongeurs
Catalog # FSCI16121-14
Supplier:  FINE SCIENCE TOOLS
Marketsource
Friedman-Pearson rongeurs
Friedman-Pearson rongeurs

Friedman-Pearson rongeurs, 0,7 mm cup diameter, Curved, 140 mm

Price
564,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These rongeurs are made of stainless steel.

  • Single joint action
Specifications
  • Type:
    Curved
  • Description:
    Friedman-Pearson rongeurs, 0,7 mm cup diameter
  • Length:
    140 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.