Order Entry
Germany
Orders LinkContactUsLinkComponent
Extra fine Bonn scissors

Extra fine Bonn scissors

Sharp/sharp, curved tips, 85 mm, 13 mm
Catalog # FSCI14085-08
Marketsource
Supplier:   FINE SCIENCE TOOLS
Extra fine Bonn scissors
Catalog # FSCI14085-08
Supplier:  FINE SCIENCE TOOLS
Marketsource
Extra fine Bonn scissors
Extra fine Bonn scissors

Sharp/sharp, curved tips, 85 mm, 13 mm

Price
151,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Sharp/sharp, curved tips
  • Length:
    85 mm
  • Seq:
    02
  • Blade length:
    13 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.