Order Entry
Germany
Orders LinkContactUsLinkComponent
Surgical scissors, blunt

Surgical scissors, blunt

Blunt/blunt, curved tips, 165 mm, 53 mm
Catalog # FSCI14003-16
Marketsource
Supplier:   FINE SCIENCE TOOLS
Surgical scissors, blunt
Catalog # FSCI14003-16
Supplier:  FINE SCIENCE TOOLS
Marketsource
Surgical scissors, blunt
Surgical scissors, blunt

Blunt/blunt, curved tips, 165 mm, 53 mm

Price
76,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Blunt/blunt, curved tips
  • Length:
    165 mm
  • Seq:
    07
  • Blade length:
    53 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.