Order Entry
Germany
Orders LinkContactUsLinkComponent
Surgical scissors, sharp-blunt

Surgical scissors, sharp-blunt

Sharp/blunt, straight tips, 145 mm, 47 mm
Catalog # FSCI14001-14
Marketsource
Supplier:   FINE SCIENCE TOOLS
Surgical scissors, sharp-blunt
Catalog # FSCI14001-14
Supplier:  FINE SCIENCE TOOLS
Marketsource
Surgical scissors, sharp-blunt
Surgical scissors, sharp-blunt

Sharp/blunt, straight tips, 145 mm, 47 mm

Price
64,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Sharp/blunt, straight tips
  • Length:
    145 mm
  • Seq:
    06
  • Blade length:
    47 mm