Surgical scissors, sharp-blunt
Sharp/blunt, curved tips, 185 mm, 57 mm
FSCI14004-18
Marketsource
These scissors are made of stainless steel.
- Type:Sharp/blunt, curved tips
- Length:185 mm
- Seq:09
- Blade length:57 mm
These scissors are made of stainless steel.