Order Entry
Germany
Orders LinkContactUsLinkComponent
Metzenbaum scissors

Metzenbaum scissors

Blunt/blunt, straight tips, 180 mm, 35 mm
Catalog # FSCI14016-18
Marketsource
Supplier:   FINE SCIENCE TOOLS
Metzenbaum scissors
Catalog # FSCI14016-18
Supplier:  FINE SCIENCE TOOLS
Marketsource
Metzenbaum scissors
Metzenbaum scissors

Blunt/blunt, straight tips, 180 mm, 35 mm

Price
99,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Blunt/blunt, straight tips
  • Length:
    180 mm
  • Seq:
    05
  • Blade length:
    35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.