Shodex GPC KF-2004 - polymer based organic HPLC column - SEC mode - THF - preparative - ID: 20mm length: 300mm - Functional Group: - - Base Material: Styrene dinvinylbenzene copolymer - Particle Size (µm): 7 - Pore Size (A): - - Exclusion Limit (g/m 1 * 1 items
SHOWF6102404
Shodex GPC KF-2004 - polymer based organic HPLC column - SEC mode - THF - preparative - ID: 20mm length: 300mm - Functional Group: - - Base Material: Styrene dinvinylbenzene copolymer - Particle Size (µm): 7 - Pore Size (A): - - Exclusion Limit (g/m 1 * 1 items
SHOWF6102404