D-Cycloserine
Glass bottle for solids, 25 g
Catalog # BACE4030155.0025
Marketsource
Supplier: BACHEM BIOCHEMICA
Jump to:
Product Details & Documents
Synonyms: D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
- CAS Number:68-41-7
- Formula:C₃H₆N₂O₂
- MDL Number:MFCD00005353
- Molecular Weight:102.09 g/mol
- Storage Temperature:Freezer
- EINECS:200-688-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



