Order Entry
Germany
Orders LinkContactUsLinkComponent
Xanthine, Sigma-Aldrich®

Xanthine, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIALX0626-5G
Irritant
Supplier:   Merck
Xanthine, Sigma-Aldrich®
Catalog # SIALX0626-5G
Supplier:  Merck
CAS Number:  69-89-6
Xanthine, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  69-89-6
Price
104,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine
  • CAS Number:69-89-6
  • Formula:C₅H₄N₄O₂
  • MDL Number:MFCD00078453
  • Molecular Weight:152.11 g/mol
  • Merck Index:12,10193
  • EINECS:200-718-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.