Order Entry
Germany
Orders LinkContactUsLinkComponent
Connectors

Connectors

Connector, 25 mm TC to 9,5 mm hose barb, Polysulfone
Catalog # KRB4-3PS
Supplier:   Cytiva
Connectors
Catalog # KRB4-3PS
Supplier:  Cytiva
Connectors

Connector, 25 mm TC to 9,5 mm hose barb, Polysulfone

Price
141,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications
  • Description:
    Connector, 25 mm TC to 9,5 mm hose barb
  • Material:
    Polysulfone
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.