Orders LinkContactUsLinkComponent
PCR cloning kit, CloneJET
PCR cloning kit, CloneJET
CloneJET PCR Cloning Kit, 40 reactions, 1 KIT
Catalog # K1232
PCR cloning kit, CloneJET
Catalog # K1232
CAS Number:  
PCR cloning kit, CloneJET
PCR cloning kit, CloneJET
CloneJET PCR Cloning Kit, 40 reactions, 1 KIT
CAS Number:  
Price
371,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
The CloneJET PCR cloning kit contains a ready-to-use positive selection cloning vector pJET1.2/blunt. The vector contains a lethal restriction enzyme gene that is disrupted by ligation of a DNA insert into the cloning site. As a result, only bacterial cells with recombinant plasmids are able to form colonies. Recircularised pJET1.2/blunt vector molecules lacking an insert express a lethal restriction enzyme, which kills the host E. coli cell after transformation. This positive selection drastically accelerates the process of colony screening and eliminates additional costs required for blue/white selection.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    CloneJET PCR Cloning Kit, 40 reactions