Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Stopcocks, straight or multi-way bore

2 mm, 4,5 mm
  BURK8610-0010
 :   BURKLE
Stopcocks, straight or multi-way bore
  BURK8610-0010
 :  BURKLE
Stopcocks, straight or multi-way bore
Stopcocks, straight or multi-way bore

2 mm, 4,5 mm

 
44,20 €
   
1 * 1 ST
 

PTFE.

  • Resistant to compression max. 1 bar
  • Vacuum suitable
  • Autoclavable
 
  • Hole Ø:
    2 mm
  • For tube I-Ø:
    4,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.