Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Bottles, serum/sample, WHEATON®

2,00 ml, 15,6×36 mm, Clear, No, 13 mm
Catalog # 548-1977
Supplier:   DWK Life Sciences
Bottles, serum/sample, WHEATON®
Catalog # 548-1977
Supplier:  DWK Life Sciences
Bottles, serum/sample, WHEATON®
Bottles, serum/sample, WHEATON®

2,00 ml, 15,6×36 mm, Clear, No, 13 mm

Price
359,00 €
Log in for availability
1 * 288 ST
Product Details & Documents

Serum bottles are ideal for long- and short-term sample storage, lyophilisation and vaccine/injectable drug containers. These moulded bottles have excellent wall strength and chemical tolerances which are important for storage applications. Bottles are manufactured from Wheaton 400 borosilicate moulded glass that conforms to USP Type I requirements. Bottles are packed in shrink-wrapped partitioned modules with the exception of 2 ml size, which has no partition.

  • Fits most freeze drying applications
  • Borosilicate glass conforms to USP Type I requirements
  • Shrink-wrapped partitioned modules reduce breakage
Specifications
  • Cap included:
    No
  • Cap Size:
    13 mm
  • Ø×H:
    15,6×36 mm
  • Capacity:
    2,00 ml
  • Colour:
    Clear
  • Seq:
    01