Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(2-Bromophenyl)ethanol

2-(2-Bromophenyl)ethanol

Glass bottle for solids, 1 g
Catalog # COBBOT-0499-1G
Marketsource
Supplier:   COMBI-BLOCKS
2-(2-Bromophenyl)ethanol
Catalog # COBBOT-0499-1G
Supplier:  COMBI-BLOCKS
Marketsource
CAS Number:  1074-16-4
2-(2-Bromophenyl)ethanol

Glass bottle for solids, 1 g

CAS Number:  1074-16-4
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:1074-16-4
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00093566
  • Molecular Weight:201.06 g/mol
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    1 g

Specifications