2-(2-Bromophenyl)ethanol
Glass bottle for solids, 5 g
Catalog # COBBOT-0499-5G
Marketsource
Supplier: COMBI-BLOCKS
2-(2-Bromophenyl)ethanol
Catalog # COBBOT-0499-5G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 1074-16-4
Jump to:
Product Details & Documents
- CAS Number:1074-16-4
- Formula:C₈H₉BrO
- MDL Number:MFCD00093566
- Molecular Weight:201.06 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



