3,4,5-Trifluorophenol
Glass bottle for solids, 5 g
Catalog # COBBOT-0593-5G
Marketsource
Supplier: COMBI-BLOCKS
3,4,5-Trifluorophenol
Catalog # COBBOT-0593-5G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 99627-05-1
Jump to:
Product Details & Documents
- CAS Number:99627-05-1
- Formula:C₆H₃F₃O
- MDL Number:MFCD00042427
- Molecular Weight:148.08 g/mol
- Density:1.629 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:436-120-9
- UN:3261
- ADR:8,II
- :Density: 1.629 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



