Order Entry
Germany
Orders LinkContactUsLinkComponent
3-Fluoropyrocatechol

3-Fluoropyrocatechol

Glass bottle for solids, 25 g
Catalog # COBBOS-7647-25G
Marketsource
Supplier:   COMBI-BLOCKS
3-Fluoropyrocatechol
Catalog # COBBOS-7647-25G
Supplier:  COMBI-BLOCKS
Marketsource
CAS Number:  363-52-0
3-Fluoropyrocatechol

Glass bottle for solids, 25 g

CAS Number:  363-52-0
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
  • CAS Number:363-52-0
  • Formula:C₆H₅FO₂
  • MDL Number:MFCD00042582
  • Molecular Weight:128.1 g/mol
  • Boiling Pt:218 °C (1013 hPa)
  • Melting Pt:71 °C
  • Density:1,514 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.