ContactUsLinkComponent
Methyl 2-amino-4-(2-chlorophenyl)thiazole-5-carboxylate, ECN [1065074-40-9], formula C11H9ClN2O2S, storage room tempratre, transport normal temperature, FW 268.7 1 * 5 g
  COBBHC-6787-5G
Methyl 2-amino-4-(2-chlorophenyl)thiazole-5-carboxylate, ECN [1065074-40-9], formula C11H9ClN2O2S, storage room tempratre, transport normal temperature, FW 268.7 1 * 5 g
  COBBHC-6787-5G