ContactUsLinkComponent
Methyl 4-cyclopropyl-2-(methylthio)pyrimidine-5-carboxylate, ECN [1072944-76-3], formula C10H12N2O2S, storage room tempratre, transp ort normal temperature, FW 224.3 1 * 5 g
Catalog # COBBHC-6783-5G
Supplier:  COMBI-BLOCKS
Methyl 4-cyclopropyl-2-(methylthio)pyrimidine-5-carboxylate, ECN [1072944-76-3], formula C10H12N2O2S, storage room tempratre, transp ort normal temperature, FW 224.3 1 * 5 g
Catalog # COBBHC-6783-5G
Supplier:  COMBI-BLOCKS