4-(Boc-amino)cyclohexanone
Glass bottle for solids, 5 g
Catalog # COBBAM-1608-5G
Marketsource
Supplier: COMBI-BLOCKS
4-(Boc-amino)cyclohexanone
Catalog # COBBAM-1608-5G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 179321-49-4
Jump to:
Product Details & Documents
Synonyms: 4-Aminocyclohexan-1-one, N-BOC protected, tert-Butyl (4-oxocyclohexyl)carbamate
- CAS Number:179321-49-4
- Formula:C₁₁H₁₉NO₃
- MDL Number:MFCD00798168
- Molecular Weight:213.2700 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



