3-(2-Chlorophenyl)propionic acid
Glass bottle for solids, 25 g
Catalog # COBBHC-2572-25G
Marketsource
Supplier: COMBI-BLOCKS
3-(2-Chlorophenyl)propionic acid
Catalog # COBBHC-2572-25G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 1643-28-3
Jump to:
Product Details & Documents
Synonyms: 3-(2-Chlorophenyl)propanoic acid
- CAS Number:1643-28-3
- Formula:C₉H₉ClO₂
- MDL Number:MFCD00016547
- Molecular Weight:184.62 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



