Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4-Pentylcyclohexanone

Glass bottle for solids, 100 g
Catalog # COBBOS-7417-100G
Marketsource
Supplier:   COMBI-BLOCKS
4-Pentylcyclohexanone
Catalog # COBBOS-7417-100G
Supplier:  COMBI-BLOCKS
Marketsource
CAS Number:  61203-83-6
4-Pentylcyclohexanone

Glass bottle for solids, 100 g

CAS Number:  61203-83-6
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:61203-83-6
  • Formula:C₁₁H₂₀O
  • MDL Number:MFCD00059581
  • Molecular Weight:168.28 g/mol
  • EINECS:406-670-4
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications