4-Pentylcyclohexanone
Glass bottle for solids, 25 g
Catalog # COBBOS-7417-25G
Marketsource
Supplier: COMBI-BLOCKS
4-Pentylcyclohexanone
Catalog # COBBOS-7417-25G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 61203-83-6
Jump to:
Product Details & Documents
- CAS Number:61203-83-6
- Formula:C₁₁H₂₀O
- MDL Number:MFCD00059581
- Molecular Weight:168.28 g/mol
- EINECS:406-670-4
- UN:3082
- ADR:9,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



