2-(4-Bromophenyl)ethanol
Glass bottle for solids, 25 g
COBBOS-7596-25G
Marketsource
: 4-Bromophenethyl alcohol
- :4654-39-1
- :C₈H₉BrO
- :MFCD00002897
- :201.06 g/mol
- :275 °C (1013 hPa)
- :36…38 °C
- :1,4678 g/cm³ (28.5 °C)
- :225-093-7
- Pack type:Glass bottle for solids
- Pk:25 g