2-(4-Bromophenyl)ethanol
Glass bottle for solids, 5 g
Catalog # COBBOS-7596-5G
Marketsource
Supplier: COMBI-BLOCKS
2-(4-Bromophenyl)ethanol
Catalog # COBBOS-7596-5G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 4654-39-1
Jump to:
Product Details & Documents
Synonyms: 4-Bromophenethyl alcohol
- CAS Number:4654-39-1
- Formula:C₈H₉BrO
- MDL Number:MFCD00002897
- Molecular Weight:201.06 g/mol
- Boiling Pt:275 °C (1013 hPa)
- Melting Pt:36…38 °C
- Density:1,4678 g/cm³ (28.5 °C)
- EINECS:225-093-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



