6-Hydroxyindole
Glass bottle for solids, 1 g
Catalog # COBBIN-0036-1G
Marketsource
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
Synonyms: Indol-6-ol, 6-Indolol, IMEXINE OBA, 6-Hydroxy-1H-indole, 1H-Indol-6-ol
- CAS Number:2380-86-1
- Formula:C₈H₇NO
- MDL Number:MFCD00152101
- Molecular Weight:133.15 g/mol
- EINECS:417-020-4
- UN:3077
- ADR:9,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



