ContactUsLinkComponent
5-Bromo-1-(4-methoxybenzyl)-1H-benzoECN [d]imidazole, ECN [1020253-11-5], formula C15H13BrN2O, storage room tempratre, transport nor mal temperature, FW 317.2 1 * 5 g
Catalog # COBBHI-1371-5G
Supplier:  COMBI-BLOCKS
5-Bromo-1-(4-methoxybenzyl)-1H-benzoECN [d]imidazole, ECN [1020253-11-5], formula C15H13BrN2O, storage room tempratre, transport nor mal temperature, FW 317.2 1 * 5 g
Catalog # COBBHI-1371-5G
Supplier:  COMBI-BLOCKS