4-Hydroxy-2,6-dimethoxybenzaldehyde
Glass bottle for solids, 1 g
Catalog # COBBBD-0035-1G
Marketsource
Supplier: COMBI-BLOCKS
4-Hydroxy-2,6-dimethoxybenzaldehyde
Catalog # COBBBD-0035-1G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 22080-96-2
Jump to:
Product Details & Documents
Synonyms: 2,6-Dimethoxy-4-hydroxybenzaldehyde
- CAS Number:22080-96-2
- Formula:C₉H₁₀O₄
- MDL Number:MFCD01407645
- Molecular Weight:182.1733 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



