Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Hydroxy-2,6-dimethoxybenzaldehyde

4-Hydroxy-2,6-dimethoxybenzaldehyde

Glass bottle for solids, 5 g
Catalog # COBBBD-0035-5G
Marketsource
Supplier:   COMBI-BLOCKS
4-Hydroxy-2,6-dimethoxybenzaldehyde
Catalog # COBBBD-0035-5G
Supplier:  COMBI-BLOCKS
Marketsource
CAS Number:  22080-96-2
4-Hydroxy-2,6-dimethoxybenzaldehyde

Glass bottle for solids, 5 g

CAS Number:  22080-96-2
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2,6-Dimethoxy-4-hydroxybenzaldehyde
  • CAS Number:22080-96-2
  • Formula:C₉H₁₀O₄
  • MDL Number:MFCD01407645
  • Molecular Weight:182.1733 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications